ADV390216988 AChemBlock

(R)-3-(Methylsulfonyl)pyrrolidine

  • MDL number MFCD17015903
  • Cas Number : 1234576-84-1
  • Molecular Weight : 149.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-3-methylsulfonyl-pyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5244
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart