ADV390216990 AChemBlock

3-(Methylsulfonyl)piperidine hydrochloride

  • MDL number MFCD17015904
  • Cas Number : 1378304-65-4
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-methylsulfonyl-piperidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5246
pictograms GHS07
hstatements H302;H313;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart