ADV390216993 AChemBlock

(S)-Pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD09025426
  • Cas Number : 1124369-40-9
  • Molecular Weight : 151.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-pyrrolidine-3-carboxylic-acid-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5250
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart