ADV390216995 AChemBlock

(S)-Methyl pyrrolidine-3-carboxylate hydrochloride

  • MDL number MFCD11840196
  • Cas Number : 1099646-61-3
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-methyl-pyrrolidine-3-carboxylate-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5252
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart