ADV390216999 AChemBlock

(S)-1-Boc-3-(hydroxymethyl)pyrrolidine

  • MDL number MFCD03094726
  • Cas Number : 199174-24-8
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless to yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-1-boc-3-hydroxymethyl-pyrrolidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5256
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart