ADV390217002 AChemBlock

(S)-Pyrrolidine-3-carboxamide hydrochloride

  • MDL number MFCD22421737
  • Cas Number : 1279048-81-5
  • Molecular Weight : 150.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-pyrrolidine-3-carboxamide-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5261
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart