ADV390217005 AChemBlock

(S)-Piperidine-3-carboxamide hydrochloride

  • MDL number MFCD22415259
  • Cas Number : 1026661-43-7
  • Molecular Weight : 164.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-piperidine-3-carboxamide-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5264
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart