ADV390217010 AChemBlock

1-Methylpyrrolidine-3-ethanol

  • MDL number MFCD17015913
  • Cas Number : 36763-96-9
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form light-yellow liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-methylpyrrolidine-3-ethanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5270
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart