ADV390217044 AChemBlock

3,3-Dimethoxycyclobutane-1-carboxylate methyl ester

  • MDL number MFCD11111944
  • Cas Number : 98231-07-3
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-3-dimethoxycyclobutane-1-carboxylate-methyl-ester.html
Availability details for
Quantity
  • Availability
  • Add to Cart