ADV390217047 AChemBlock

Ethyl 2-Bromo-3-methoxy-6-pyridinecarboxylate

  • MDL number MFCD14698338
  • Cas Number : 1214329-41-5
  • Molecular Weight : 260.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-bromo-3-methoxy-6-pyridinecarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4405
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart