ADV390217066 AChemBlock

4-Chloro-5-fluoro-2-(methylsulfanyl)pyrimidine

  • MDL number MFCD14636507
  • Cas Number : 6096-45-3
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-5-fluoro-2-methylsulfanyl-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5307
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart