ADV390217071 AChemBlock

5-Chloro-2-pyrazinemethanol

  • MDL number MFCD11110224
  • Cas Number : 72788-94-4
  • Molecular Weight : 144.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-2-pyrazinemethanol.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5313
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart