ADV390217075 AChemBlock

5-Hydroxymethyl-1-methyl-1H-pyridin-2-one

  • MDL number MFCD17015927
  • Cas Number : 27330-18-3
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-hydroxymethyl-1-methyl-1h-pyridin-2-one.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5318
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart