ADV390217100 AChemBlock

(S)-3-Iodo-tetrahydrofuran

  • MDL number MFCD17015958
  • Cas Number : 918439-76-6
  • Molecular Weight : 198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-3-iodo-tetrahydrofuran.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5349
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart