ADV390217118 AChemBlock

Ethyl 7-chlorothieno[3,2-b]pyridine-6-carboxylate

  • MDL number MFCD08234993
  • Cas Number : 83179-01-5
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-7-chlorothieno-3-2-b-pyridine-6-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5385
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart