ADV390217120 AChemBlock

2-Chloro-7-iodo-5H-pyrrolo[2,3-b]pyrazine

  • MDL number MFCD12755891
  • Cas Number : 889447-20-5
  • Molecular Weight : 279.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-7-iodo-5h-pyrrolo-2-3-b-pyrazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5387
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart