ADV390217124 AChemBlock

[1,2,4]Triazolo[1,5-a]pyridine-5-carboxylic acid

  • MDL number MFCD17015980
  • Cas Number : 1234616-38-6
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-4-triazolo-1-5-a-pyridine-5-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5392
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart