ADV390217131 AChemBlock

3-Chloropyrazine-2-carboxylic acid

  • MDL number MFCD01318438
  • Cas Number : 27398-39-6
  • Molecular Weight : 158.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form brown powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-chloropyrazine-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5403
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart