ADV390217132 AChemBlock

3-Amino-6-chloropyridine-2-carboxylic acid hydrochloride

  • MDL number MFCD19689878
  • Cas Number : 1112213-20-3
  • Molecular Weight : 209.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-6-chloropyridine-2-carboxylic-acid-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5404
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart