ADV390217150 AChemBlock

5-Bromo-4-chloro-2,3-diaminopyridine

  • MDL number MFCD11865186
  • Cas Number : 1131604-99-3
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form orange solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-chloro-2-3-diaminopyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5427
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart