ADV390217171 AChemBlock

8-Bromo-2-chloroquinazoline

  • MDL number MFCD11043845
  • Cas Number : 956100-63-3
  • Molecular Weight : 243.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-bromo-2-chloroquinazoline.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5448
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart