ADV390217175 AChemBlock

3-Methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD11857900
  • Cas Number : 1118787-14-6
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-1h-pyrazolo-3-4-b-pyridine-5-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5452
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart