ADV390217187 AChemBlock

6-Bromo-1,3-dimethyl-1H-indazole

  • MDL number MFCD12028626
  • Cas Number : 1095539-84-6
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-1-3-dimethyl-1h-indazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5464
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart