ADV390217196 AChemBlock

Methyl 2-chloroquinazoline-8-carboxylate

  • MDL number MFCD15072116
  • Cas Number : 1217269-81-2
  • Molecular Weight : 222.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-chloroquinazoline-8-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5474
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart