ADV390217207 AChemBlock

2-Methyl-1,5-naphthyridine

  • MDL number MFCD07437992
  • Cas Number : 7675-32-3
  • Molecular Weight : 144.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pink solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-1-5-naphthyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5488
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart