ADV390217212 AChemBlock

5-Bromo-2,2-difluoro-benzo[1,3]dioxole

  • MDL number MFCD00236212
  • Cas Number : 33070-32-5
  • Molecular Weight : 237

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-2-difluoro-benzo-1-3-dioxole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5493
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart