ADV390217220 AChemBlock

6-amino-2,2-dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one

  • MDL number MFCD10000812
  • Cas Number : 1002726-62-6
  • Molecular Weight : 193.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2h-pyrido-3-2-b-1-4-oxazin-3-4h-one-6-amino-2-2-dimethyl.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5501
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart