ADV390217224 AChemBlock

2,4-Dichloro-5-methyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD13189357
  • Cas Number : 1060815-86-2
  • Molecular Weight : 202.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-5-methyl-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5506
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart