ADV390217230 AChemBlock

7-Chloro-2-iodothieno[3,2-b]pyridine

  • MDL number MFCD08448166
  • Cas Number : 602303-26-4
  • Molecular Weight : 295.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-chloro-2-iodothieno-3-2-b-pyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5512
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart