ADV390217235 AChemBlock

2,6-Dichloro-4-methyl-3-nitropyridine

  • MDL number MFCD11110694
  • Cas Number : 60010-03-9
  • Molecular Weight : 207.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-6-dichloro-4-methyl-3-nitropyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5517
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart