ADV390217238 AChemBlock

5-Chloro-3-thiophenemethanamine hydrochloride

  • MDL number MFCD17016065
  • Cas Number : 1108712-55-5
  • Molecular Weight : 184.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3-thiophenemethanamine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5520
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart