ADV390217243 AChemBlock

5-Boc-4,6-dihydro-1H-pyrrolo[3,4-c]pyrazole

  • MDL number MFCD08061257
  • Cas Number : 657428-42-7
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-boc-4-6-dihydro-1h-pyrrolo-3-4-c-pyrazole.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5526
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart