ADV390217247 AChemBlock

Ethyl 2-amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxylate

  • MDL number MFCD05664038
  • Cas Number : 117642-16-7
  • Molecular Weight : 227.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-amino-4-7-dihydro-5h-thieno-2-3-c-pyran-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5531
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart