ADV390217249 AChemBlock

Ethyl 5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate hydrochloride

  • MDL number MFCD09259960
  • Cas Number : 723286-68-8
  • Molecular Weight : 232.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-6-7-8-tetrahydro-1-2-4-triazolo-4-3-a-pyrazine-3-carboxylate-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5533
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart