ADV390217259 AChemBlock

1-Boc-3-ethynylpyrrolidine

  • MDL number MFCD10698160
  • Cas Number : 287193-00-4
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-ethynylpyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5545
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart