ADV390217266 AChemBlock

1H-Imidazole-5-acetonitrile

  • MDL number MFCD05863381
  • Cas Number : 18502-05-1
  • Molecular Weight : 107.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-imidazole-5-acetonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5552
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart