ADV390217284 AChemBlock

Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate

  • MDL number MFCD10566741
  • Cas Number : 342613-63-2
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white to cream solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-bromoimidazo-1-2-a-pyridine-7-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5574
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart