ADV390217301 AChemBlock

3,4-Dichloro-5-nitropyridine

  • MDL number MFCD09910258
  • Cas Number : 56809-84-8
  • Molecular Weight : 192.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form orange solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-4-dichloro-5-nitropyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5594
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart