ADV390217316 AChemBlock

7-Amino-3-bromo-1-methyl-1H-indazole

  • MDL number MFCD08234922
  • Cas Number : 885271-76-1
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-amino-3-bromo-1-methyl-1h-indazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5610
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart