ADV390217327 AChemBlock

5-Cyano-2-fluorothiophene

  • MDL number MFCD11042555
  • Cas Number : 32415-91-1
  • Molecular Weight : 127.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-cyano-2-fluorothiophene.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5622
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart