ADV390217335 AChemBlock

4-Chloro-7-methylthieno[3,2-d]pyrimidine

  • MDL number MFCD00205202
  • Cas Number : 175137-21-0
  • Molecular Weight : 184.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7-methylthieno-3-2-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5630
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart