ADV390217336 AChemBlock

3-Amino-6-iodopyridazine

  • MDL number MFCD09991754
  • Cas Number : 187973-60-0
  • Molecular Weight : 221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pale yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-6-iodopyridazine.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3343
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart