ADV390217337 AChemBlock

4-Amino-5-bromo-2-methoxypyridine

  • MDL number MFCD12025502
  • Cas Number : 1232431-48-9
  • Molecular Weight : 203.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-5-bromo-2-methoxypyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5633
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart