ADV390217348 AChemBlock

cis-4-Amino-1-Boc-3-methoxypiperidine

  • MDL number MFCD09749826
  • Cas Number : 1171124-68-7
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cis-4-amino-1-boc-3-methoxypiperidine.html

Safety Information

signalword Warning
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5653
pictograms GHS05
hstatements H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart