ADV390217387 AChemBlock

3,3-Difluorocyclobutanecarbonitrile

  • MDL number MFCD10001482
  • Cas Number : 86770-80-1
  • Molecular Weight : 117.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluorocyclobutanecarbonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5726
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart