ADV390217399 AChemBlock

4-Amino-2-(methylthio)-5-pyrimidinecarboxaldehyde

  • MDL number MFCD08669438
  • Cas Number : 770-31-0
  • Molecular Weight : 169.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-2-methylthio-5-pyrimidinecarboxaldehyde.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5751
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart