ADV390217406 AChemBlock

6-Bromo-1,4-dichlorophthalazine

  • MDL number MFCD16037285
  • Cas Number : 240400-95-7
  • Molecular Weight : 277.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Store at Room Tem.
Physical Form white solid
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/6-bromo-1-4-dichlorophthalazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3398
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart