ADV390217413 AChemBlock

4-Bromo-3-ethylpyridine hydrobromide

  • MDL number MFCD23106325
  • Cas Number : 1523606-24-7
  • Molecular Weight : 266.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-3-ethylpyridine-hydrobromide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5778
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart