ADV390217417 AChemBlock

3,4-Dinitro-1H-pyrazole

  • MDL number MFCD00159604
  • Cas Number : 38858-92-3
  • Molecular Weight : 158.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-4-dinitro-1h-pyrazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5783
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart