ADV390217423 AChemBlock

(3R)-2,2-Dimethyl-thiomorpholine-3-carboxylic acid

  • MDL number MFCD08063022
  • Cas Number : 774243-35-5
  • Molecular Weight : 175.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3r-2-2-dimethyl-thiomorpholine-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5796
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart